AMBH580649D5 Ambeed, Inc.

6-(Trifluoromethyl)pyridine-2,3-dicarboxylic acid

  • MDL number MFCD22576273
  • Cas Number : 90376-94-6
  • Molecular Weight : 235.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-(trifluoromethyl)pyridine-2,3-dicarboxylic acid
Inchi Code 1S/C8H4F3NO4/c9-8(10,11)4-2-1-3(6(13)14)5(12-4)7(15)16/h1-2H,(H,13,14)(H,15,16)
Inchi Key LXJKAVDDRGVIQD-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/90376-94-6.html

Safety Information

signalword Warning
pcodes P305+P351+P338-P280
MSDS https://www.ambeed.com/products/90376-94-6.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart