AMBH580649D9 Ambeed, Inc.

1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-�D-glucopyranose

  • MDL number MFCD00080781
  • Cas Number : 10022-13-6
  • Molecular Weight : 477.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/10022-13-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/10022-13-6.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart