AMBH580649DF Ambeed, Inc.

Methyl 6-chloro-2,4-dimethylnicotinate

  • MDL number MFCD18258696
  • Cas Number : 1256789-09-9
  • Molecular Weight : 199.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1256789-09-9.html

Safety Information

MSDS https://www.ambeed.com/products/1256789-09-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart