AMBH580649E0 Ambeed, Inc.

2-Hydroxybenzene-1,3,5-tricarbaldehyde

  • MDL number MFCD01314207
  • Cas Number : 81502-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/81502-74-1.html

Safety Information

MSDS https://www.ambeed.com/products/81502-74-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart