AMBH580649E5 Ambeed, Inc.

4-Amino-1-(1-boc-azetidin-3-yl)-1H-pyrazole

  • MDL number MFCD10687122
  • Cas Number : 1029413-51-1
  • Molecular Weight : 238.2862

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form Pale-yellow to Yellow-brown to Brown to Red Liquid or Solid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1029413-51-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1029413-51-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart