AMBH580649E9 Ambeed, Inc.

2,5,6-Trichlorobenzo[d]thiazole

  • MDL number MFCD09749269
  • Cas Number : 120258-61-9
  • Molecular Weight : 238.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8
Country of Origin China
Product Link https://www.ambeed.com/products/120258-61-9.html

Safety Information

MSDS https://www.ambeed.com/products/120258-61-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart