AMBH580649EC Ambeed, Inc.

2-((tert-Butoxycarbonyl)amino)-2-(tetrahydrofuran-3-yl)acetic acid

  • MDL number MFCD04115582
  • Cas Number : 874583-03-6
  • Molecular Weight : 245.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/874583-03-6.html

Safety Information

MSDS https://www.ambeed.com/products/874583-03-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart