AMBH580649EE Ambeed, Inc.

GNE-317

  • MDL number MFCD28900677
  • Cas Number : 1394076-92-6
  • Molecular Weight : 414.4814

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/1394076-92-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1394076-92-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart