AMBH580649F5 Ambeed, Inc.

2-Methyl-3-(trifluoromethyl)benzonitrile

  • MDL number MFCD01631593
  • Cas Number : 261952-02-7
  • Molecular Weight : 185.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/261952-02-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/261952-02-7.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart