AMBH580649F7 Ambeed, Inc.

(E)-1-Pentene-1-boronic Acid Pinacol Ester

  • MDL number MFCD05663882
  • Cas Number : 161395-96-6
  • Molecular Weight : 196.0942

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Physical Form Solid or liquid
Shipping Temperature Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/161395-96-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/161395-96-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart