AMBH58064A05 Ambeed, Inc.

4,4',4'',4'''-(Ethene-1,1,2,2-tetrayl)tetrabenzaldehyde

  • MDL number MFCD31700764
  • Cas Number : 2170451-48-4
  • Molecular Weight : 444.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/2170451-48-4.html

Safety Information

MSDS https://www.ambeed.com/products/2170451-48-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart