AMBH58064A06 Ambeed, Inc.

(2E)-3-Phenyl-1-(pyridin-2-yl)prop-2-en-1-one

  • MDL number MFCD00223896
  • Cas Number : 53940-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/53940-12-8.html

Safety Information

MSDS https://www.ambeed.com/products/53940-12-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart