AMBH58064A07 Ambeed, Inc.

GSK369796 Dihydrochloride

  • MDL number MFCD16627991
  • Cas Number : 1010411-21-8
  • Molecular Weight : 428.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-[(tert-butylamino)methyl]-5-[(7-chloro-4-quinolinyl)amino]phenol dihydrochloride
Inchi Code 1S/C20H22ClN3O.2ClH/c1-20(2,3)23-12-13-4-6-15(11-19(13)25)24-17-8-9-22-18-10-14(21)5-7-16(17)18;;/h4-11,23,25H,12H2,1-3H3,(H,22,24);2*1H
Inchi Key UDVALKJFXQVZSI-UHFFFAOYSA-N

Description

Synonym 2-((tert-Butylamino)methyl)-5-((7-chloroquinolin-4-yl)amino)phenol dihydrochloride
Purity 98%
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1010411-21-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1010411-21-8.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart