AMBH58064A0B Ambeed, Inc.

4-(3-Tosyl-2-(tosylmethyl)propanoyl)benzoic acid

  • MDL number MFCD28142486
  • Cas Number : 124243-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/124243-00-1.html

Safety Information

MSDS https://www.ambeed.com/products/124243-00-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart