AMBH58064A0E Ambeed, Inc.

5-Chloro-4-fluoro-2-iodoaniline

  • MDL number MFCD27929959
  • Cas Number : 852569-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/852569-38-1.html

Safety Information

MSDS https://www.ambeed.com/products/852569-38-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart