AMBH58064A17 Ambeed, Inc.

5-Amino-2-bromo-3-fluoropyridine

  • MDL number MFCD21099397
  • Cas Number : 1256276-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/1256276-41-1.html

Safety Information

MSDS https://www.ambeed.com/products/1256276-41-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart