AMBH58064A19 Ambeed, Inc.

N-Methyl-N-nitroguanidine

  • MDL number MFCD00024167
  • Cas Number : 4245-76-5
  • Molecular Weight : 118.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-155°C at N/A mmHg
Purity 99
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/4245-76-5.html

Safety Information

signalword Danger
pcodes P210-P261-P280-P305+P351+P338
hcodes 4.1
MSDS https://www.ambeed.com/products/4245-76-5.html
pictograms GHS07,GHS02
hstatements H228-H302-H312-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart