AMBH58064A21 Ambeed, Inc.

(2S,4R)-Methyl 4-fluoropyrrolidine-2-carboxylate hydrochloride

  • MDL number MFCD07374354
  • Cas Number : 58281-80-4
  • Molecular Weight : 183.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/58281-80-4.html

Safety Information

MSDS https://www.ambeed.com/products/58281-80-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart