AMBH5816B59F Ambeed, Inc.

6-Methoxy-3,3-dimethyl-3,4-dihydroisoquinolin-1(2H)-one

  • MDL number MFCD19349280
  • Cas Number : 1267456-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/1267456-79-0.html

Safety Information

MSDS https://www.ambeed.com/products/1267456-79-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart