AMBH5816B5F5 Ambeed, Inc.

(S)-5-Chloro-2,3-dihydro-1H-inden-1-amine hydrochloride

  • MDL number MFCD22375542
  • Cas Number : 1376687-76-1
  • Molecular Weight : 204.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1376687-76-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1376687-76-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart