AMBH5816B60A Ambeed, Inc.

(2R,3R,4S,5R)-2-Bromotetrahydro-2H-pyran-3,4,5-triyl triacetate

  • MDL number MFCD00070174
  • Cas Number : 3068-31-3
  • Molecular Weight : 339.1400

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/3068-31-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/3068-31-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart