AMBH5816B620 Ambeed, Inc.

tert-Butyl 5-bromo-3-methyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

  • MDL number MFCD18260123
  • Cas Number : 1111637-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1111637-66-1.html

Safety Information

MSDS https://www.ambeed.com/products/1111637-66-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart