AMBH5816B621 Ambeed, Inc.

4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxamide

  • MDL number N/A
  • Cas Number : 182808-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/182808-16-8.html

Safety Information

MSDS https://www.ambeed.com/products/182808-16-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart