AMBH5816B627 Ambeed, Inc.

2-Acetyloxazole-4-carboxylic acid

  • MDL number MFCD26679540
  • Cas Number : 1297542-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1297542-52-9.html

Safety Information

MSDS https://www.ambeed.com/products/1297542-52-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart