AMBH5816B62D Ambeed, Inc.

(S)-6-(4-Chlorophenyl)-2-(3-fluorophenyl)-N-(1-hydroxypropan-2-yl)-3-oxo-2,3-dihydropyridazine-4-carboxamide

  • MDL number MFCD31736303
  • Cas Number : 2162982-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/2162982-11-6.html

Safety Information

MSDS https://www.ambeed.com/products/2162982-11-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart