AMBH5816B62E Ambeed, Inc.

N-(4-(Methylamino)phenyl)acetamide hydrochloride

  • MDL number N/A

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/P001060863.html

Safety Information

MSDS https://www.ambeed.com/products/P001060863.html
Availability details for
Quantity
  • Availability
  • Add to Cart