AMBH5816B637 Ambeed, Inc.

2,3-Dimethylpyrido[3,4-b]pyrazin-5(6H)-one

  • MDL number N/A

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/P001063884.html

Safety Information

MSDS https://www.ambeed.com/products/P001063884.html
Availability details for
Quantity
  • Availability
  • Add to Cart