AMBH5816B638 Ambeed, Inc.

Methyl 2-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)acetate

  • MDL number MFCD08753504
  • Cas Number : 25379-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/25379-89-9.html

Safety Information

MSDS https://www.ambeed.com/products/25379-89-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart