AMBH5816B69D Ambeed, Inc.

3-(4-(Methylthio)phenyl)acrylic acid

  • MDL number MFCD00266583
  • Cas Number : 102016-58-0
  • Molecular Weight : 194.2500

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/102016-58-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/102016-58-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart