AMBH5816B728 Ambeed, Inc.

3,4-Dicaffeoylquinic Acid

  • MDL number MFCD08272439
  • Cas Number : 14534-61-3
  • Molecular Weight : 516.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/14534-61-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/14534-61-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart