AMBH5816B883 Ambeed, Inc.

3-Bromothieno[2,3-c]pyridine

  • MDL number MFCD11052627
  • Cas Number : 28783-17-7
  • Molecular Weight : 214.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 145.9°C
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 317.6°C at 760 mmHg
Product Link https://www.ambeed.com/products/28783-17-7.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/28783-17-7.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart