AMBH5816B893 Ambeed, Inc.

1-(4-Methoxypyrimidin-2-yl)ethanone

  • MDL number MFCD24712087
  • Cas Number : 463337-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/463337-53-3.html

Safety Information

MSDS https://www.ambeed.com/products/463337-53-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart