AMBH5816B90D Ambeed, Inc.

GSK-2018682

  • MDL number N/A
  • Cas Number : 1034688-30-6
  • Molecular Weight : 440.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(4-(5-(5-Chloro-6-isopropoxypyridin-3-yl)-1,2,4-oxadiazol-3-yl)-1H-indol-1-yl)butanoic acid
Purity 97
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1034688-30-6.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/1034688-30-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart