AMBH5816B946 Ambeed, Inc.

5-Bromo-1H-pyrrole-2-carbonitrile

  • MDL number MFCD23135667
  • Cas Number : 36953-44-3
  • Molecular Weight : 170.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20°C
Physical Form Solid or liquid
Shipping Temperature Shipped in cold pack
Country of Origin China
Product Link https://www.ambeed.com/products/36953-44-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/36953-44-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart