AMBH5816B97F Ambeed, Inc.

3-Bromofuro[2,3-b]pyridine

  • MDL number MFCD19687765
  • Cas Number : 92404-58-5
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-95°C
Purity 95
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 247.6°20.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/92404-58-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/92404-58-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart