AMBH581765E4 Ambeed, Inc.

5-Methyl-3,4-dihydroquinolin-2(1H)-one

  • MDL number MFCD13179101
  • Cas Number : 20151-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/20151-46-6.html

Safety Information

MSDS https://www.ambeed.com/products/20151-46-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart