AMBH581765EB Ambeed, Inc.

3-Bromo-6-chloro-1-methylpyridin-2(1H)-one

  • MDL number MFCD21603985
  • Cas Number : 960299-32-5
  • Molecular Weight : 222.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/960299-32-5.html

Safety Information

MSDS https://www.ambeed.com/products/960299-32-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart