AMBH581765EF Ambeed, Inc.

4-Methylbenzyl acetate

  • MDL number MFCD00017234
  • Cas Number : 2216-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/2216-45-7.html

Safety Information

MSDS https://www.ambeed.com/products/2216-45-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart