AMBH581765F1 Ambeed, Inc.

1-(Difluoromethyl)-3-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD20330757
  • Cas Number : 1339559-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/1339559-21-5.html

Safety Information

MSDS https://www.ambeed.com/products/1339559-21-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart