AMBH581765FA Ambeed, Inc.

[1,2,4]Triazolo[4,3-a]pyrimidin-3(2H)-one

  • MDL number MFCD13193348
  • Cas Number : 63206-77-9
  • Molecular Weight : 136.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/63206-77-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/63206-77-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart