AMBH581765FF Ambeed, Inc.

2-Amino-3-ethylbenzoic acid

  • MDL number MFCD11193650
  • Cas Number : 5437-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/5437-40-1.html

Safety Information

MSDS https://www.ambeed.com/products/5437-40-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart