AMBH58176601 Ambeed, Inc.

Direct Red 28

  • MDL number MFCD00004028
  • Cas Number : 573-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://www.ambeed.com/products/573-58-0.html

Safety Information

MSDS https://www.ambeed.com/products/573-58-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart