AMBH5817660A Ambeed, Inc.

Guanosine 5'-phosphate disodium salt hydrate

  • MDL number N/A
  • Cas Number : 146894-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/146894-16-8.html

Safety Information

MSDS https://www.ambeed.com/products/146894-16-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart