AMBH5817660C Ambeed, Inc.

4-(2-(1-(p-Tolyl)ethylidene)hydrazinyl)benzenesulfonamide

  • MDL number N/A
  • Cas Number : 1061214-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1061214-06-9.html

Safety Information

MSDS https://www.ambeed.com/products/1061214-06-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart