AMBH5817660D Ambeed, Inc.

(2-Oxo-2,3-dihydrobenzo[d]oxazol-5-yl)boronic acid

  • MDL number MFCD22572000
  • Cas Number : 710348-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/710348-42-8.html

Safety Information

MSDS https://www.ambeed.com/products/710348-42-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart