AMBH5817660E Ambeed, Inc.

2-(((6-(Difluoromethoxy)-1H-benzo[d]imidazol-2-yl)thio)methyl)-3,4-dimethoxypyridine 1-oxide

  • MDL number MFCD18252639
  • Cas Number : 953787-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin China
Product Link https://www.ambeed.com/products/953787-51-4.html

Safety Information

MSDS https://www.ambeed.com/products/953787-51-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart