AMBH58176613 Ambeed, Inc.

SB-423562

  • MDL number MFCD19443794
  • Cas Number : 351490-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/351490-27-2.html

Safety Information

MSDS https://www.ambeed.com/products/351490-27-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart