AMBH58176614 Ambeed, Inc.

(1S,2R)-2-Aminocyclobutanol hydrochloride

  • MDL number MFCD19215995
  • Cas Number : 206751-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/206751-79-3.html

Safety Information

MSDS https://www.ambeed.com/products/206751-79-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart