AMBH58176616 Ambeed, Inc.

4,6-Dichlorothieno[2,3-d]pyrimidine

  • MDL number MFCD27937516
  • Cas Number : 1638764-42-7
  • Molecular Weight : 205.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/1638764-42-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1638764-42-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart