AMBH5817662B Ambeed, Inc.

2-Amino-3-methylbenzaldehyde

  • MDL number MFCD18389804
  • Cas Number : 84902-24-9
  • Molecular Weight : 135.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/84902-24-9.html

Safety Information

MSDS https://www.ambeed.com/products/84902-24-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart